Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
ENST00000312527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
ENST00000392580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
ENST00000537389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029685.hg.1 |
TC11002475.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012930.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000312527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000392580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000524765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
ENST00000531778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
NM_018644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
NM_054025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199064.hg.1 |
TC1100012930.hg.1 |
Clariom D TC |