Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014494.2 | NP_055309.2 | 13 - 14 | 4103 | 81 | |
XM_005255257.4 | XP_005255314.1 | 12 - 13 | 4070 | 81 | ||
XM_017023143.1 | XP_016878632.1 | 12 - 13 | 4047 | 81 | ||
XM_017023144.1 | XP_016878633.1 | 11 - 12 | 4014 | 81 | ||
XM_017023145.1 | XP_016878634.1 | 12 - 13 | 4049 | 81 | ||
XM_017023146.1 | XP_016878635.1 | 12 - 13 | 4112 | 81 | ||
XM_017023148.1 | XP_016878637.1 | 11 - 12 | 3867 | 81 | ||
XM_017023152.1 | XP_016878641.1 | 11 - 12 | 3619 | 81 | ||
XM_017023153.1 | XP_016878642.1 | 12 - 13 | 4070 | 81 | ||
XM_017023154.1 | XP_016878643.1 | 10 - 11 | 3961 | 81 | ||
GenBank mRNA | AB040893.1 | - | 8 - 9 | 2303 | 81 | |
AJ492221.1 | - | 12 - 13 | 3989 | 81 | ||
AY035864.1 | - | 10 - 11 | 3637 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |