Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013866.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013866.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013866.hg.1 |
TC15001476.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000558694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
ENST00000566386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009522.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170284.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170284.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170284.hg.1 |
ENST00000260453.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170284.hg.1 |
NM_018365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170284.hg.1 |
TC1500009522.hg.1 |
Clariom D TC |