Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000378803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000395035.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000419111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000419111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000419111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000419111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000451199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000451199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000451199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
ENST00000451199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012318.hg.1 |
NM_001009565.2 |
Refseq |