Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005868.hg.1 |
TC0X000446.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005868.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005868.hg.1 |
ENST00000373119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005868.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005868.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153250.hg.1 |
TC0X00007798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373114.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
ENST00000373119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_019117.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007798.hg.1 |
NM_057162.2 |
Refseq |