Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008957.hg.1 |
TC05000647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008957.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008957.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008957.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008955.hg.1 |
TC05000647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008955.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008955.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008955.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159391.hg.1 |
TC0500008654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159389.hg.1 |
TC0500008654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159388.hg.1 |
TC0500008654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000296877.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000483190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
ENST00000485457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008654.hg.1 |
NM_052971.2 |
Refseq |