Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023072.hg.1 |
TC08001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023072.hg.1 |
ENST00000307404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023072.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
ENST00000616126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012202.hg.1 |
NR_107057.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000563210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000568812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000569712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
ENST00000616126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012202.hg.1 |
NR_107057.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183468.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183468.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183468.hg.1 |
ENST00000569669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183468.hg.1 |
NM_015201.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183468.hg.1 |
TC0800012202.hg.1 |
Clariom D TC |