Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015246.hg.1 |
TC07001002.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015246.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015246.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015248.hg.1 |
TC07001002.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015248.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015248.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015247.hg.1 |
TC07001002.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015247.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07015247.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168708.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168708.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168708.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168708.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168708.hg.1 |
TC0700009675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700168710.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168710.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168710.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168710.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168710.hg.1 |
TC0700009675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700168709.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168709.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168709.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168709.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168709.hg.1 |
TC0700009675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700168707.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168707.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168707.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168707.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168707.hg.1 |
TC0700009675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |