Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
TC02000524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
ENST00000312912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
ENST00000409064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
ENST00000409556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
ENST00000540058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
ENST00000542128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
NM_001146688.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008185.hg.1 |
NM_018433.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000312912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000409064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000409556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000441719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000462197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000542128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
NM_001146688.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
NM_018433.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000312912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000409064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000409556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000441719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000462197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
ENST00000542128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
NM_001146688.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
NM_018433.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157634.hg.1 |
TC0200008291.hg.1 |
Clariom D TC |