Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018605.hg.1 |
ENST00000372202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018605.hg.1 |
ENST00000372204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018605.hg.1 |
ENST00000421924.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018605.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018605.hg.1 |
TC10001463.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176466.hg.1 |
TC1000011215.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000372204.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000421924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000453477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
ENST00000454362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_001269053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011215.hg.1 |
NM_138812.3 |
Refseq |