Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017027205.1 | XP_016882694.1 | 2 - 3 | 2193 | 65 | |
GenBank mRNA | AK127948.1 | - | 2 - 3 | 282 | 65 | |
DA476988.1 | - | 2 - 3 | 284 | 65 | ||
DA746547.1 | - | 2 - 3 | 286 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |