Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007656.hg.1 |
ENST00000519514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000341298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
ENST00000368400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_018973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015943.hg.1 |
NM_153741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007656.hg.1 |
ENST00000519514.1 |
Ensembl |