Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199282.2 | NP_001186211.2 | 32 - 33 | 5671 | 76 | |
NM_006726.4 | NP_006717.2 | 32 - 33 | 5630 | 76 | ||
XM_005263372.3 | XP_005263429.1 | 32 - 33 | 5816 | 76 | ||
XM_005263373.2 | XP_005263430.1 | 32 - 33 | 5858 | 76 | ||
XM_005263374.3 | XP_005263431.1 | 32 - 33 | 5815 | 76 | ||
XM_005263375.3 | XP_005263432.1 | 32 - 33 | 5815 | 76 | ||
XM_011532434.2 | XP_011530736.1 | 32 - 33 | 5815 | 76 | ||
XM_017008872.1 | XP_016864361.1 | 32 - 33 | 5858 | 76 | ||
GenBank mRNA | AB208838.1 | - | 8 - 9 | 1344 | 76 | |
AF216648.2 | - | 32 - 33 | 5630 | 76 | ||
AF467287.1 | - | 32 - 33 | 5671 | 76 | ||
M83822.1 | - | 14 - 15 | 3066 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |