Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015885.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219363.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219363.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219363.hg.1 |
ENST00000368655.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219363.hg.1 |
NM_020699.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219363.hg.1 |
TC0100015885.hg.1 |
Clariom D TC |