Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195454.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195454.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195454.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500195454.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500195454.hg.1 |
TC0500013300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000263408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000467285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
ENST00000509186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013300.hg.1 |
NM_001737.4 |
Refseq |