Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024550.1 | 8 - 9 | 1003 | 79 | ||
XM_017012871.1 | XP_016868360.1 | 9 - 10 | 2019 | 79 | ||
XM_017012872.1 | XP_016868361.1 | 8 - 9 | 1925 | 79 | ||
XM_017012873.1 | XP_016868362.1 | 9 - 10 | 1814 | 79 | ||
XM_017012874.1 | XP_016868363.1 | 9 - 10 | 1050 | 79 | ||
XM_017012877.1 | XP_016868366.1 | 8 - 9 | 912 | 79 | ||
GenBank mRNA | AF202144.1 | - | 8 - 9 | 952 | 79 | |
AK126664.1 | - | 8 - 9 | 1498 | 79 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927420 | LOC101927420 | -22.9kb | Sense | Chr. 7 - 87151423 - 87152493 on NCBI Build GRCh38 | |
Downstream | transmembrane protein 243 | TMEM243 |
C7orf23, MM-TRAG, MMTRAG |
14.8kb | Sense | Chr. 7 - 87195593 - 87220603 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |