Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005607.hg.1 |
TC04000405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005607.hg.1 |
ENST00000226355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005607.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153645.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153645.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153645.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153645.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153645.hg.1 |
TC0400007832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000226355.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
ENST00000505794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007832.hg.1 |
NM_001133.2 |
Refseq |