Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142535.1 | NP_001136007.1 | 6 - 7 | 975 | 105 | |
NM_001142537.1 | NP_001136009.1 | 5 - 6 | 1062 | 105 | ||
NM_033101.3 | NP_149092.2 | 6 - 7 | 972 | 105 | ||
XM_017018458.1 | XP_016873947.1 | 6 - 7 | 975 | 105 | ||
GenBank mRNA | AF222694.1 | - | 5 - 6 | 654 | 105 | |
AF222695.1 | - | 5 - 6 | 542 | 105 | ||
AF244974.1 | - | 6 - 7 | 941 | 105 | ||
AF244975.1 | - | 5 - 6 | 1062 | 105 | ||
AF310686.1 | - | 6 - 7 | 586 | 105 | ||
AF310687.1 | - | 6 - 7 | 589 | 105 | ||
AK313851.1 | - | 6 - 7 | 972 | 105 | ||
BC028222.1 | - | 6 - 7 | 922 | 5217281 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |