Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015888.4 | NP_056972.1 | 17 - 18 | 1918 | 77 | |
XM_011541562.2 | XP_011539864.1 | 17 - 18 | 1888 | 77 | ||
GenBank mRNA | AF044923.1 | - | 17 - 18 | 1860 | 77 | |
AK292314.1 | - | 18 - 19 | 1763 | 77 | ||
AK301768.1 | - | 17 - 18 | 1686 | 77 | ||
BC011621.2 | - | 17 - 18 | 1918 | 3959931 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |