Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023729.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023734.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023734.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023734.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023734.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023734.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023736.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023736.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023736.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023736.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023730.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023731.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023731.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023731.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023731.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023731.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023738.hg.1 |
ENST00000279488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023738.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023738.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023738.hg.1 |
TC12001796.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188838.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188843.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188842.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188850.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188844.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188841.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188847.hg.1 |
TC1200011470.hg.1 |
Clariom D TC |