Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036.4 | NP_001027.3 | 33 - 34 | 4474 | 84 | |
NM_001243996.2 | NP_001230925.1 | 33 - 34 | 4474 | 84 | ||
XM_011521880.2 | XP_011520182.1 | 33 - 34 | 4662 | 84 | ||
XM_017022468.1 | XP_016877957.1 | 33 - 34 | 4662 | 84 | ||
XM_017022469.1 | XP_016877958.1 | 33 - 34 | 4662 | 84 | ||
XM_017022470.1 | XP_016877959.1 | 33 - 34 | 6996 | 84 | ||
XM_017022471.1 | XP_016877960.1 | 33 - 34 | 4659 | 84 | ||
XM_017022472.1 | XP_016877961.1 | 33 - 34 | 4662 | 84 | ||
XM_017022473.1 | XP_016877962.1 | 33 - 34 | 4662 | 84 | ||
XM_017022474.1 | XP_016877963.1 | 33 - 34 | 4626 | 84 | ||
XM_017022475.1 | XP_016877964.1 | 33 - 34 | 4662 | 84 | ||
XM_017022476.1 | XP_016877965.1 | 32 - 33 | 4554 | 84 | ||
XM_017022477.1 | XP_016877966.1 | 33 - 34 | 4663 | 84 | ||
GenBank mRNA | AB001025.1 | - | 33 - 34 | 4479 | 84 | |
AJ001515.1 | - | 33 - 34 | 4460 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |