Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026889.hg.1 |
ENST00000288532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026889.hg.1 |
ENST00000302223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026889.hg.1 |
ENST00000445328.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026889.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026889.hg.1 |
TC12002047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026890.hg.1 |
ENST00000288532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026890.hg.1 |
ENST00000302223.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026890.hg.1 |
ENST00000445328.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026890.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026890.hg.1 |
TC12002047.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194907.hg.1 |
TC1200012167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000546838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000547943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012167.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000288532.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000445328.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000547736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000551769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
ENST00000552443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
NM_032314.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194903.hg.1 |
TC1200012167.hg.1 |
Clariom D TC |