Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016425.hg.1 |
TC08001275.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016425.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016425.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016425.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016424.hg.1 |
TC08001275.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016424.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016424.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172278.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172278.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172278.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172278.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172278.hg.1 |
TC0800010648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010648.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
ENST00000523954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172279.hg.1 |
TC0800010648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172277.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172277.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172277.hg.1 |
ENST00000310193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172277.hg.1 |
NM_004820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172277.hg.1 |
TC0800010648.hg.1 |
Clariom D TC |