Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
TC03001688.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
ENST00000264235.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
ENST00000539838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030454.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000473886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
ENST00000474830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012132.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
ENST00000264235.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
ENST00000316626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
NM_001146156.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
NM_002093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190314.hg.1 |
TC0300012132.hg.1 |
Clariom D TC |