Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004877.3 | NP_004868.1 | 2 - 3 | 206 | 64 | |
GenBank mRNA | AB001993.1 | - | 1 - 2 | 106 | 64 | |
AF038956.1 | - | 1 - 2 | 100 | 64 | ||
BC022207.1 | - | 2 - 3 | 126 | 64 | ||
BC032819.2 | - | 1 - 2 | 93 | 64 | ||
BC080180.1 | - | 2 - 3 | 126 | 5419511 | 64 | |
BC093799.1 | - | 2 - 3 | 159 | 7939644 | 64 | |
BC101818.1 | - | 2 - 3 | 159 | 8069324 | 64 | |
BC143543.1 | - | 2 - 3 | 159 | 9052054 | 64 | |
BC143546.1 | - | 2 - 3 | 158 | 9052057 | 64 | |
BC143548.1 | - | 2 - 3 | 157 | 9052059 | 64 | |
BF974742.1 | - | 2 - 3 | 207 | 64 | ||
CR456967.1 | - | 1 - 2 | 96 | 64 | ||
CR542114.1 | - | 1 - 2 | 96 | 64 | ||
EB387540.1 | - | 2 - 3 | 305 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |