Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
TC02001649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
ENST00000355123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
ENST00000361999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
ENST00000380868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
ENST00000445614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
NM_006277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02026280.hg.1 |
NM_019595.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000355123.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000361999.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000622089.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
NM_006277.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
NM_019595.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000355123.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000361999.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
ENST00000622089.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
NM_006277.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
NM_019595.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187057.hg.1 |
TC0200011980.hg.1 |
Clariom D TC |