Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01052270_m1
See other ABI2 GE Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01052270_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001282925.1
NP_001269854.1
4 - 5
759
66
XM_005246217.2
XP_005246274.1
4 - 5
1124
66
XM_006712174.2
XP_006712237.1
4 - 5
1124
66
XM_006712177.2
XP_006712240.1
5 - 6
1200
66
XM_006712179.2
XP_006712242.1
4 - 5
1124
66
XM_006712180.2
XP_006712243.1
4 - 5
1124
66
XM_006712181.2
XP_006712244.1
4 - 5
1124
66
XM_006712182.2
XP_006712245.1
4 - 5
1124
66
XM_011510472.2
XP_011508774.1
4 - 5
1124
66
XM_017003118.1
XP_016858607.1
4 - 5
1124
66
XM_017003119.1
XP_016858608.1
4 - 5
1124
66
XM_017003120.1
XP_016858609.1
4 - 5
1124
66
XM_017003121.1
XP_016858610.1
4 - 5
1124
66
XM_017003122.1
XP_016858611.1
4 - 5
1124
66
XM_017003128.1
XP_016858617.1
5 - 6
1200
66
XM_017003129.1
XP_016858618.1
4 - 5
1124
66
XM_017003130.1
XP_016858619.1
4 - 5
1124
66
XM_017003132.1
XP_016858621.1
4 - 5
1124
66
XM_017003133.1
XP_016858622.1
5 - 6
1200
66
XM_017003137.1
XP_016858626.1
5 - 6
1200
66
XM_017003138.1
XP_016858627.1
4 - 5
1124
66
XM_017003143.1
XP_016858632.1
5 - 6
1200
66
XM_017003144.1
XP_016858633.1
4 - 5
1124
66
GenBank mRNA
AF260261.1
-
4 - 5
483
66
AK299092.1
-
2 - 3
43
66
AK316000.1
-
6 - 7
698
66
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the AK299092.1 transcripts.
Set Membership:
Amplicon length less than or equal to 70
Homo sapiens
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0