Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045154.hg.1 |
TC01002892.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045154.hg.1 |
ENST00000370205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045154.hg.1 |
ENST00000507727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045154.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045155.hg.1 |
TC01002892.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045155.hg.1 |
ENST00000370205.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045155.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211824.hg.1 |
TC0100014997.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000495856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014997.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
ENST00000370205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
ENST00000507727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NM_001305242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NM_144988.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
NR_131032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211823.hg.1 |
TC0100014997.hg.1 |
Clariom D TC |