Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000521085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
ENST00000521085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
TC08000629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
ENST00000251808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
ENST00000395928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
ENST00000517674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008616.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157882.hg.1 |
TC0800008416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000251808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000395927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000472106.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000474338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000517674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000521085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
ENST00000521085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008416.hg.1 |
NM_024915.3 |
Refseq |