Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010103.hg.1 |
TC01000593.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010103.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010103.hg.1 |
ENST00000538609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010103.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158553.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158553.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158553.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158553.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158553.hg.1 |
TC0100008182.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000371901.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
ENST00000466294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008182.hg.1 |
NM_178033.1 |
Refseq |