Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027442.hg.1 |
TC03001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027442.hg.1 |
ENST00000358781.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027442.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027436.hg.1 |
TC03001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027436.hg.1 |
ENST00000358781.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027436.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027435.hg.1 |
TC03001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027435.hg.1 |
ENST00000358781.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027435.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027441.hg.1 |
TC03001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027441.hg.1 |
ENST00000358781.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027441.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184943.hg.1 |
TC0300011313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184937.hg.1 |
TC0300011313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000482623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011313.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000483787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
ENST00000498347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184935.hg.1 |
TC0300011313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
ENST00000358781.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
ENST00000489857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
NM_138805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184944.hg.1 |
TC0300011313.hg.1 |
Clariom D TC |