Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034104.hg.1 |
TC01002205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034104.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034104.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034106.hg.1 |
TC01002205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034106.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034106.hg.1 |
ENST00000474547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034106.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034105.hg.1 |
TC01002205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034105.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034105.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034103.hg.1 |
TC01002205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034103.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034103.hg.1 |
ENST00000474547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034103.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195339.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195339.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195339.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195339.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195339.hg.1 |
TC0100012898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012898.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195337.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195337.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195337.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195337.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195337.hg.1 |
TC0100012898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100195338.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195338.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195338.hg.1 |
ENST00000376468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100195338.hg.1 |
NM_002521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100195338.hg.1 |
TC0100012898.hg.1 |
Clariom D TC |