Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000567834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008210.hg.1 |
ENST00000307892.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008210.hg.1 |
ENST00000512062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008210.hg.1 |
ENST00000515314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008210.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008210.hg.1 |
TC16000569.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161857.hg.1 |
TC1600008228.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161856.hg.1 |
TC1600008228.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000514123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000561792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000567834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
ENST00000568342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008228.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000307892.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000512062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000515314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
ENST00000568237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
NM_030579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161855.hg.1 |
TC1600008228.hg.1 |
Clariom D TC |