Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024690.2 | NP_078966.2 | 71 - 72 | 42357 | 72 | |
XM_017027486.1 | XP_016882975.1 | 65 - 66 | 41437 | 72 | ||
XM_017027487.1 | XP_016882976.1 | 64 - 65 | 41371 | 72 | ||
XM_017027489.1 | XP_016882978.1 | 62 - 63 | 41283 | 72 | ||
XM_017027490.1 | XP_016882979.1 | 60 - 61 | 40972 | 72 | ||
XM_017027491.1 | XP_016882980.1 | 60 - 61 | 40972 | 72 | ||
XM_017027492.1 | XP_016882981.1 | 60 - 61 | 40969 | 72 | ||
XM_017027493.1 | XP_016882982.1 | 60 - 61 | 40969 | 72 | ||
XM_017027494.1 | XP_016882983.1 | 60 - 61 | 40969 | 72 | ||
XM_017027495.1 | XP_016882984.1 | 60 - 61 | 40969 | 72 | ||
XM_017027496.1 | XP_016882985.1 | 45 - 46 | 39568 | 72 | ||
XM_017027497.1 | XP_016882986.1 | 40 - 41 | 39094 | 72 | ||
XM_017027499.1 | XP_016882988.1 | 65 - 66 | 32224 | 72 | ||
XM_017027500.1 | XP_016882989.1 | 64 - 65 | 32143 | 72 | ||
XM_017027501.1 | XP_016882990.1 | 44 - 45 | 4151 | 72 | ||
GenBank mRNA | AF361486.3 | - | 19617 | 72 | ||
AK128857.1 | - | 3987 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |