Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314019.1 | NP_001300948.1 | 4 - 5 | 286 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |