Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000918.hg.1 |
ENST00000270139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000918.hg.1 |
ENST00000416947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000918.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000918.hg.1 |
TC21000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000493503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
ENST00000493503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000442071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000493503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
ENST00000493503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006960.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146750.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146750.hg.1 |
NM_000629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146750.hg.1 |
TC2100006960.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100146750.hg.1 |
ENST00000270139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146750.hg.1 |
NM_000629.2 |
Refseq |