Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004815.3 | NP_004806.3 | 17 - 18 | 2295 | 102 | |
XM_006711048.3 | XP_006711111.1 | 16 - 17 | 1929 | 102 | ||
XM_011542438.2 | XP_011540740.1 | 17 - 18 | 2170 | 102 | ||
XM_011542439.2 | XP_011540741.1 | 17 - 18 | 2015 | 102 | ||
XM_011542440.2 | XP_011540742.1 | 16 - 17 | 1889 | 102 | ||
GenBank mRNA | AB208873.1 | - | 16 - 17 | 1901 | 102 | |
AK307733.1 | - | 17 - 18 | 2124 | 102 | ||
AK310491.1 | - | 17 - 18 | 2015 | 102 | ||
AL833445.1 | - | 5 - 6 | 6504 | 102 | ||
BC093741.1 | - | 16 - 17 | 1968 | 7939586 | 102 | |
BC093767.1 | - | 16 - 17 | 1971 | 7939612 | 102 | |
U90920.1 | - | 17 - 18 | 2109 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |