Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000282753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000355289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000361719.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000492807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000502405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
ENST00000507907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009740.hg.1 |
NM_001278067.1 |
Refseq |