Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145913.3 | NP_666018.3 | 10 - 11 | 1624 | 72 | |
XM_017018910.1 | XP_016874399.1 | 10 - 11 | 1660 | 72 | ||
GenBank mRNA | AF536216.1 | - | 10 - 11 | 1597 | 72 | |
AK075263.1 | - | 10 - 11 | 1282 | 72 | ||
AK313788.1 | - | 10 - 11 | 1593 | 72 | ||
AY081220.1 | - | 10 - 11 | 1598 | 72 | ||
BC110492.2 | - | 10 - 11 | 1259 | 40007567 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |