Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000402938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
ENST00000602808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012540.hg.1 |
NM_002043.4 |
Refseq |