Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005000.hg.1 |
TC04000369.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005000.hg.1 |
ENST00000305363.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005000.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR4_ctg9_hap1000004.hg.1 |
TC4_ctg9_hap1000001.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR4_ctg9_hap1000004.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007734.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
ENST00000305363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
ENST00000510647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
NM_014058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152696.hg.1 |
TC0400007734.hg.1 |
Clariom D TC |