Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000767.hg.1 |
TC08000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000767.hg.1 |
ENST00000221086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000767.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000767.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000768.hg.1 |
TC08000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000768.hg.1 |
ENST00000221086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000768.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000768.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000528389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006738.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145632.hg.1 |
TC0800006738.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000221086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000526292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
ENST00000530200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
NM_015458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145630.hg.1 |
TC0800006738.hg.1 |
Clariom D TC |