Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043190.hg.1 |
TC01002765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043190.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043190.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014542.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208592.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208592.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208592.hg.1 |
ENST00000262340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208592.hg.1 |
NM_000329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208592.hg.1 |
TC0100014542.hg.1 |
Clariom D TC |