Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004830.hg.1 |
TC03000239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004830.hg.1 |
ENST00000396056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004830.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000296092.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000399560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
ENST00000453164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007216.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000396056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000415571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
ENST00000432115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
NM_003420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150936.hg.1 |
TC0300007216.hg.1 |
Clariom D TC |