Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
ENST00000338916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
ENST00000544769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023822.hg.1 |
TC19001751.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
TC1900012009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900207399.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000338916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000593901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000595557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000598997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
ENST00000600079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_001160329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012009.hg.1 |
NM_032298.2 |
Refseq |