Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000720.3 | NP_000711.1 | 26 - 27 | 3344 | 74 | |
NM_001128839.2 | NP_001122311.1 | 25 - 26 | 3284 | 74 | ||
NM_001128840.2 | NP_001122312.1 | 25 - 26 | 3284 | 74 | ||
XM_005265448.3 | XP_005265505.1 | 25 - 26 | 3166 | 74 | ||
XM_011534094.2 | XP_011532396.1 | 26 - 27 | 3352 | 74 | ||
XM_011534096.2 | XP_011532398.1 | 25 - 26 | 3292 | 74 | ||
XM_011534097.2 | XP_011532399.1 | 24 - 25 | 3819 | 74 | ||
XM_011534099.2 | XP_011532401.1 | 22 - 23 | 2802 | 74 | ||
XM_011534100.2 | XP_011532402.1 | 25 - 26 | 3292 | 74 | ||
XM_017007137.1 | XP_016862626.1 | 26 - 27 | 3352 | 74 | ||
XM_017007138.1 | XP_016862627.1 | 26 - 27 | 3349 | 74 | ||
XM_017007139.1 | XP_016862628.1 | 26 - 27 | 3352 | 74 | ||
XM_017007140.1 | XP_016862629.1 | 25 - 26 | 3292 | 74 | ||
XM_017007141.1 | XP_016862630.1 | 25 - 26 | 3292 | 74 | ||
XM_017007142.1 | XP_016862631.1 | 26 - 27 | 3352 | 74 | ||
XM_017007143.1 | XP_016862632.1 | 26 - 27 | 3352 | 74 | ||
XM_017007144.1 | XP_016862633.1 | 26 - 27 | 3352 | 74 | ||
XM_017007145.1 | XP_016862634.1 | 26 - 27 | 3352 | 74 | ||
GenBank mRNA | AB209171.1 | - | 20 - 21 | 2245 | 74 | |
EU363339.1 | - | 25 - 26 | 3166 | 74 | ||
M76558.1 | - | 25 - 26 | 3676 | 74 | ||
M83566.1 | - | 26 - 27 | 3344 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Homo sapiens Probe spans exons |