Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001280.hg.1 |
TC03000056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001280.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001280.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145664.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145664.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145664.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145664.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145664.hg.1 |
TC0300006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
ENST00000256458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006580.hg.1 |
NM_001570.3 |
Refseq |