Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
ENST00000373744.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
ENST00000433211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
ENST00000540598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
ENST00000545309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016872.hg.1 |
TC10001338.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173714.hg.1 |
TC1000010818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173715.hg.1 |
TC1000010818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000330298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000373735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000433211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000494580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
ENST00000545309.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001127384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_001291133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010818.hg.1 |
NM_013266.3 |
Refseq |