Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006454.hg.1 |
TC01000421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006454.hg.1 |
ENST00000373602.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006454.hg.1 |
ENST00000421922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006454.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153853.hg.1 |
TC0100007669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000461819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000469003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
ENST00000483009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007669.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
ENST00000373602.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
ENST00000421922.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
NM_001301011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
NM_024296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153852.hg.1 |
TC0100007669.hg.1 |
Clariom D TC |