Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145064.2 | NP_001138536.1 | 9 | 108 | ||
XM_017012574.1 | XP_016868063.1 | 8 | 108 | |||
GenBank mRNA | BC048422.1 | - | 1 | 108 | ||
CR743064.1 | - | 1 | 159 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |